missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | VCX |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18696035
|
Novus Biologicals
NBP2-46770-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654516
|
Novus Biologicals
NBP2-46770 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
VCX Polyclonal antibody specifically detects VCX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Spezifikation
| VCX | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| J3KNW2 | |
| 26609 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| Variable charge X-linked protein 1;VCX1;VCX10R;VCX-10r;VCXB1;VCX-B1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MSPKPRASGPPAKATEAGKRKSSSQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts