missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46770
This item is not returnable.
View return policy
Description
VCX Polyclonal antibody specifically detects VCX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| VCX | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| J3KNW2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 26609 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Variable charge X-linked protein 1;VCX1;VCX10R;VCX-10r;VCXB1;VCX-B1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MSPKPRASGPPAKATEAGKRKSSSQ | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction