missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1/CD106 Mouse anti-Rat, Clone: MR106, Novus Biologicals™
Mouse Monoclonal Antibody
£140.00
Specifications
| Antigen | VCAM-1/CD106 |
|---|---|
| Clone | MR106 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry |
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| VCAM-1/CD106 | |
| 0.5 mg/mL | |
| Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Rat | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| VCAM1 | |
| IgG1 κ | |
| Affinity Purified |
| MR106 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 7412 | |
| The immunogen for this antibody was CD106. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title