missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1/CD106 Mouse anti-Rat, Clone: MR106, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43368-0.025MG
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Rat | |
| IgG1 κ |
| Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry | |
| MR106 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| VCAM1 | |
| The immunogen for this antibody was CD106. | |
| 0.025 mg | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 7412 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction