missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vav (pan) Antibody (SR2313), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | Vav (pan) |
|---|---|
| Clone | SR2313 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616084
|
Novus Biologicals
NBP3-22658-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624504
|
Novus Biologicals
NBP3-22658-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vav (pan) Monoclonal antibody specifically detects Vav (pan) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
| Vav (pan) | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| IgG | |
| Affinity purified |
| SR2313 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| A synthesized peptide derived from human Vav (pan) (Uniprot #: P15498, P52735, Q9UKW4) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title