missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vav (pan) Antibody (SR2313), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22658-100ul
This item is not returnable.
View return policy
Description
Vav (pan) Monoclonal antibody specifically detects Vav (pan) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Specifications
| Vav (pan) | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| A synthesized peptide derived from human Vav (pan) (Uniprot #: P15498, P52735, Q9UKW4) | |
| 100 μg | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| SR2313 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction