missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP, p Ser238 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£194.00 - £316.00
Specifications
| Antigen | VASP (p Ser238) |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18034886
|
Novus Biologicals
NB100-82254-0.025ML |
0.025 mL |
£194.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18241510
|
Novus Biologicals
NB100-82254 |
0.05 mL |
£316.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VASP Polyclonal specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VASP (p Ser238) | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| P50552 | |
| 7408 | |
| The antiserum was produced against synthesized phosphopeptide derived from human VASP around the phosphorylation site of serine 238 (K-V-SP-K-Q). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Phospho Specific | |
| vasodilator-stimulated phosphoprotein | |
| VASP | |
| IgG | |
| Affinity Purified | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title