missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP, p Ser238 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82254
This item is not returnable.
View return policy
Description
VASP Polyclonal specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| VASP (p Ser238) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| vasodilator-stimulated phosphoprotein | |
| Rabbit | |
| 50 kDa | |
| 0.05 mL | |
| Phospho Specific | |
| 7408 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P50552 | |
| VASP | |
| The antiserum was produced against synthesized phosphopeptide derived from human VASP around the phosphorylation site of serine 238 (K-V-SP-K-Q). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of VASP only when phosphorylated at serine 238. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction