missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VASA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | VASA |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL, Immunohistochemistry : 2-5 μg/mL at pH 6 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066830
|
Novus Biologicals™
NBP3-30963-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067013
|
Novus Biologicals™
NBP3-30963-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VASA Polyclonal antibody specifically detects VASA in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| VASA | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 4, DEAD box protein 4, EC 3.6.1, EC 3.6.4.13, MGC111074, probable ATP-dependent RNA helicase DDX4, Vasa homolog, VASADEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4 | |
| Synthetic peptide derived from the human VASA protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1-5 μg/mL, Immunohistochemistry : 2-5 μg/mL at pH 6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 54514 | |
| IgG | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title