missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VASA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30963-100ug
This item is not returnable.
View return policy
Description
VASA Polyclonal antibody specifically detects VASA in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| VASA | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL, Immunohistochemistry : 2-5 μg/mL at pH 6 | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 4, DEAD box protein 4, EC 3.6.1, EC 3.6.4.13, MGC111074, probable ATP-dependent RNA helicase DDX4, Vasa homolog, VASADEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4 | |
| Synthetic peptide derived from the human VASA protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 54514 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction