missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Varitip P Tips
GREENER_CHOICE

Product Code. 10493891 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
1 to 10 mL
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10493891

Brand: Eppendorf™ 0030048130

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these tips to meet a variety of needs for different lab applications. Eppendorf™ Varitip P Tips are designed for aspirating 1 to 10mL from vessels such as beakers.

  • A continuously adjustable pipettor for precise pipetting of volumes up to 10mL in 10μL increments
  • Even liquids with a high vapor pressure or viscosity can be pipetted with ease
  • System consists of a dispensing part and the Maxitip air displacement principle
  • No carry over as Maxitip exchanged
  • Valve for Maxitip ensures drip free dispensing of liquids with a high vapor pressure
  • Graduated Maxitip G for compensation of nominal value of Varipette™
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 10 mL
Filtered Non-filtered
For Use With (Application) Aspirating 2.5mL to 10mL from tall, narrow neck vessels
Tip Style Universal Fit
Graduated Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.