missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Varioskan™ LUX multimode microplate reader

Product Code. 17179510 Shop All Thermo Scientific Products
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17179510

Brand: Thermo Scientific™ VLB000GD1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Denna artikel kan inte returneras. Se returpolicy

Analyze a variety of samples with five measurement technologies, including absorbance (UV-Vis), fluorescence intensity, Luminescence, AlphaScreen / AlphaLISA and TRF.

Designed for bioscience researchers with a wide variety of needs, the Thermo Scientific™ Varioskan™ LUX multimode microplate reader comes equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. The instrument simplifies measurement setup with the automatic dynamic range selection, and its smart safety controls ease workflow and help avoid experimental errors. Varioskan LUX raises the bar for reliability and ease, with technology that works in researchers' best interest.

The Varioskan LUX multimode reader has received HTRF™ certification from Cisbio Bioassays and DLReady™ validation from Promega Corporation.

The Varioskan LUX is a modular, upgradable instrument that provides excellent usability across a variety of applications.

  • Up to five measurement technologies: Absorbance (UV-Vis), Fluorescence intensity (incl. FRET), Luminescence (direct and filtered), AlphaScreen/AlphaLISA and Time-resolved fluorescence (incl. TR-FRET/hTRF)
  • Filter and monochromator optics
  • Spectral scanning for assay optimization
  • Five measurement modes including end point, kinetic, spectra, kinetic spectra and multipoint
  • Integrated gas control module for setting CO2 and O2 concentrations for cell based assays
  • Simultaneous dispensing and measurement for signal monitoring at the start of a reaction
  • Automatic dynamic range selection, based on signal intensity of each well
  • Smart safety controls designed to identify and alert the user of potential problems before they happen
  • Automated instrument calibration and self-diagnostics for reliable performance
  • Certified HTRF compatible reader by CisBio Bioassays
  • Validated DLReady by Promega Corporation

Varioskan LUX is paired with the powerful and intuitive SkanIt Software, providing excellent usability and flexibility.

  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting Tool makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there's no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
    • Export data in a single mouse click, or have it emailed automatically at the end of the run
    • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt
  • Shows real-time spectra and kinetic curves during the measurement
  • Built-in calculations for data analysis (e.g. blank subtraction, average/SD/CV%, standard curve, dose response, quality control, classification, kinetic and spectral analysis, custom formula, etc.)
  • Comprehensive result reports with traceability to runtime actions and settings
  • Two editions available: Research Edition for life science researchers, and Drug Discovery Edition which offers features needed for the FDA 21 CFR Part 11 compliance.

The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.

TRUSTED_SUSTAINABILITY

Specifikationer

Compatibility Robot
Detector Type Photomultiplier Tube (PMT)
Incubator Temperature 45°C
Light Source Xenon Flash Lamp
Measurement Speed 96 wells: 15 sec., 384 wells: 45 sec., 1536 wells: 135 sec.
Measurement Technology Absorbance, Fluorescence Intensity
Plate Type 6 to 1536-well Plates (Absorbance 6 to 384-well Plates)
Product Line Varioskan
Reading Options Top and Bottom
Read-out Range 0 to 6 Abs
Shaking Orbital
Software Type SkanIt Software
Spectral Scanning Speed < 2.2 sec./well 400 to 500 nm, 1 flash, 2 nm Step
Wavelength Selection Double monochromaters, Double excitation monochromaters, Double emission monochromators
Accuracy (Photometric) 0.003 Abs or ±2% at 200 to 399 nm (0 to 2 Abs), 0.003 Abs or ±1%, at 400 to 1000 nm (0 to 3 Abs)
CO2 Concentration Range 0.1 to 15%
Depth (English) 23 in.
Depth (Metric) 58 cm
Dynamic Range Fluorescence intensity: top reading > 6 decades
Height (English) 20 in.
Height (Metric) 51 cm
Linearity 96-well plate: 0 to 4 Abs, ±2% at 450 nm, 384-well plate: 0 to 3 Abs ±2% at 450 nm
Measurement Type Endpoint, Kinetic, Spectra, Multipoint, Kinetic Spectra
No. of Dispensers 1
O2 Concentration Range 1 to 21%
Precision SD <0.001 Abs or CV <0.5%, at 450 nm (0 to 3 Abs)
Sensitivity Fluorescence intensity (top reading): < 0.4 fmol fluorescein/well (black 384-well plate)
Software Language Versions English, German, French, Spanish, Portuguese, Italian, Japansese, Chinese and Russian
Wavelength Range Absorbance: 200 to 1000 nm, Fluorescence intensity: Excitation: 200 to 1000 nm, Emission: 270 to 840 nm
Weight (English) 125 lb.
Weight (Metric) 57 kg
Width (English) 21 in.
Width (Metric) 53 cm
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.