missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Varian Splitless, Slotted Liners

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan™ 092228

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12954071

  • £550.00 / Pack of 25
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Varian instruments
Specifications

Specifications

GC Inlet Liner, Splitless Liner with Slots at Both Ends
74 mm
6.3 mm
25/Pk.
2 mm
Varian 1075/1077
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers