missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP-1/AOC3 Antibody (TK8-14) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £481.00
Specifications
| Antigen | VAP-1/AOC3 |
|---|---|
| Clone | TK8-14 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18291723
|
Novus Biologicals
NBP2-62556-0.025MG |
0.025 mg |
£222.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609037
|
Novus Biologicals
NBP2-62556 |
0.2 mg |
£481.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.Specifications
| VAP-1/AOC3 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| amine oxidase, copper containing 3 (vascular adhesion protein 1), Copper amine oxidase, EC 1.4.3, HPAOSSAO, Semicarbazide-sensitive amine oxidase, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1 | |
| AOC3 | |
| IgG2a κ | |
| Recognises human Vascular Adhesion Protein-1 (VAP-1), a glycosylated homodimeric membrane protein consisting of two 90 kDa subunits connected by disulfide bonds. Epitope recognized by TK8-18 is present both in dimeric and monomeric forms of VAP-1. Elevated soluble VAP-1 serum- levels have been described in several inflammatory- diseases as well as colorectal cancer. TK8-14 is a function-blocking mAb. |
| TK8-14 | |
| Monoclonal | |
| Purified | |
| PBS | |
| 8639 | |
| Affinity purified VAP-1 from tonsil stroma of human origin. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title