missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP-1/AOC3 Antibody (TK8-14) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62556
This item is not returnable.
View return policy
Description
VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.
Specifications
| VAP-1/AOC3 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| AOC3 | |
| Affinity purified VAP-1 from tonsil stroma of human origin. | |
| 8639 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| TK8-14 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence | |
| amine oxidase, copper containing 3 (vascular adhesion protein 1), Copper amine oxidase, EC 1.4.3, HPAOSSAO, Semicarbazide-sensitive amine oxidase, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1 | |
| Mouse | |
| 0.2 mg | |
| Recognises human Vascular Adhesion Protein-1 (VAP-1), a glycosylated homodimeric membrane protein consisting of two 90 kDa subunits connected by disulfide bonds. Epitope recognized by TK8-18 is present both in dimeric and monomeric forms of VAP-1. Elevated soluble VAP-1 serum- levels have been described in several inflammatory- diseases as well as colorectal cancer. TK8-14 is a function-blocking mAb. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction