missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vang-like Protein 2/VANGL2 [p Thr78, p Ser79] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33567-20ul
This item is not returnable.
View return policy
Description
Vang-like Protein 2/VANGL2 Monoclonal antibody specifically detects Vang-like Protein 2/VANGL2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Vang-like Protein 2/VANGL2 | |
| Monoclonal | |
| Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL | |
| KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2 | |
| A phospho specific peptide corresponding to residues surrounding T78/S79 of human Vang-like Protein 2/VANGL2 | |
| 20 μL | |
| Cardiovascular Biology, Neuroscience, Stem Cells | |
| 57216 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction