missing translation for 'onlineSavingsMsg'
Learn More

Vang-like Protein 2/VANGL2 [p Thr78, p Ser79] Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£161.00 - £383.00

Specifications

Antigen Vang-like Protein 2/VANGL2
Dilution Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233038
View Documents
Novus Biologicals
NBP3-33567-100ul
100 μL
£383.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229053
View Documents
Novus Biologicals
NBP3-33567-20ul
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vang-like Protein 2/VANGL2 Monoclonal antibody specifically detects Vang-like Protein 2/VANGL2 in Human samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Vang-like Protein 2/VANGL2
ELISA, Western Blot
Unconjugated
Rabbit
Cardiovascular Biology, Neuroscience, Stem Cells
PBS (pH 7.3), 50% glycerol, 0.05% BSA
57216
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
Human
KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2
A phospho specific peptide corresponding to residues surrounding T78/S79 of human Vang-like Protein 2/VANGL2
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.