missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride, AcroSeal™

Product Code. 11986171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986171 100 mL 100mL
11996171 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986171 Supplier Thermo Scientific Acros Supplier No. 433661000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19559-06-9 | VCl3·C12H24O3 | 373.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19559-06-9 , 75-09-2
Molecular Formula VCl3·C12H24O3
Molecular Weight (g/mol) 373.62
InChI Key WARDLUZTYLPMGJ-UHFFFAOYSA-K
Synonym trichlorotris tetrahydrofuran vanadium, tetrahydrofuran; trichlorovanadium, vanadium iii chloride tetrahydrofuran adduct, vanadium iii chloride tetrahydrofuran complex
PubChem CID 71311352
IUPAC Name oxolane;trichlorovanadium
SMILES C1CCOC1.Cl[V](Cl)Cl

Specifications

Color Red
Physical Form Solution
Quantity 100 mL
Linear Formula VCl·3THF
Flash Point −8°C
Formula Weight 373.62
Concentration or Composition (by Analyte or Components) 0.40 to 0.60M
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.312
Density 1.3120g/mL
Chemical Name or Material Vanadium(III) chloride-tetrahydrofuran complex (1:3)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.