accessibility menu, dialog, popup

UserName

CAS RN 19559-06-9

OClVClCl

CAS RN 19559-06-9

IUPAC Name: oxolane; trichlorovanadium
Synonyms: THF; trichlorovanadium
Molecular Weight (g/mol): 229.4
Molecular Formula: C4H8Cl3OV
InChi Key: WARDLUZTYLPMGJ-UHFFFAOYSA-K
SMILES: Cl[V](Cl)Cl.C1CCOC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results