missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10738-1-AP-150UL
This item is not returnable.
View return policy
Description
The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer′s disease, VAMP levels of all isoforms appear to be significantly lowered.Specifications
| VAMP4 | |
| Polyclonal | |
| Unconjugated | |
| VAMP4 | |
| VAMP 4, VAMP24, VAMP4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 364033, 53330, 8674 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70480, O75379 | |
| Vamp4 | |
| VAMP4 Fusion Protein Ag1090 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction