missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VAMP-2 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | VAMP-2 |
|---|---|
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066933
|
Novus Biologicals™
NBP3-30983-250ug |
250 μg |
£386.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066989
|
Novus Biologicals™
NBP3-30983-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAMP-2 Polyclonal antibody specifically detects VAMP-2 in Human,Rat samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| VAMP-2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2) | |
| Full length VAMP-2 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Sheep | |
| Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Phosphate buffered saline | |
| 6844 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title