Learn More
Novus Biologicals™ VAMP-2 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30983-50ug
 View more versions of this product
Product Code. 30066989
  • £162.00 / 50µg
Description
VAMP-2 Polyclonal antibody specifically detects VAMP-2 in Human,Rat samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAMP-2
Polyclonal
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Human, Rat
Purified
Immunoprecipitation, Western Blot
Unconjugated
FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2)
Full length VAMP-2
50 μg
Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
6844
Store at -20°C. Avoid freeze/thaw cycles.