Learn More
DWK Life Sciences DURAN™ Vacuum Receiver Adapter, bent, with 2 standard ground joints, and screw-on plastic hose connection
Product Code. 10385191 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
NS 14/23
NS 24/29
NS 29/32
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size unitSize
10385191 NS 29/32 Each
10069940 NS 14/23 Each
10633792 NS 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10385191 Supplier DWK Life Sciences Supplier No. 241304603
Only null left
Add to basket
Add to basket
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask.
  • Tight closure due to standard ground joint
  • Bent
  • With two standard ground joint and screw-on plastic hose connection
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Description Autoclavable
For Use With (Application) Distillation
Fits Tubing ID (Metric) 8.6 mm
Material Borosilicate Glass (3.3), PBT, Polypropylene, Silicone
Quantity 1 Ea.
Thread Size NS 29/32