missing translation for 'onlineSavingsMsg'
Learn More

Welch™ DuoSeal™ Vacuum Pumps

Product Code. 11553485 Shop All Welch Ilmvac Products
Change view
Click to view available options
Plug Type:
Continental European Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
11553485 Continental European Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11553485 Supplier Welch™ Supplier No. F1402C02

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Electrical Requirements 220 V, 50 Hz
Dimensions (L x W x H) 20 x 12 x 15 in.
Plug Type Continental European Plug
Weight (English) 112 lb.
No. of Stages Two
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.