Learn More
Gardner Denver Welch™ Kit 7 Vacuum pump measurement package with PIZA 111 or Gold, DN 25 KF
Shop All Welch Ilmvac Products
Click to view available options
Size:
DN 25 KF
Unit Size:
Each
Description
Vacuum measurement package consists of:
- PIZA111cr-GOLD handheld vacuum gauge
- KF25/16 measuring T-piece
- Stand for PIZA gauge
- Clamps and centring rings
Specifications
Specifications
| For Use With | Rotary vane pumps with KF25 flanges |
| Size | DN 25 KF |
| Type | Vacuum Measurement Kit |