Learn More
BrandTech™ VACUUBRAND™ VACUUBUS™ CVC3000 Vacuum Controller Solenoid Valves
For use with CVC3000 Vacuum Controller
Brand:  VACUUBRAND™ 20674217
242.20 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10099052
  • £294.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • State-of-the-art vacuum gauges for laboratory and industrial operation have to meet high standards in terms of gauge head resistance to chemicals, reliability in rugged applications and operator-friendly interfaces. VACUUBRAND has a comprehensive range of electronic measuring instruments for the range of atmospheric pressure to 5 x 10-9 mbar.
  • by cyclically turning the pump on/off (with CVC 3000 controller with module VMS-B)
  • by cyclically opening/closing an in-line solenoid valve (CVC 3000) or
  • by changing the pump´s motor speed continuously (VARIO™ pumps in connection with CVC 3000)
Specifications
Specifications
CVC3000 vacuum controller
Solenoid Vent Valve