Learn More
VACUUBRAND™ VACUU·SELECT Vacuum Controller
GREENER_CHOICE
Product Code. 16251752 Shop All Vacuubrand Products
Click to view available options
Limitations:
0.1 to 1080 mbar
Unit Size:
Each
Product Code. 16251752
Brand: VACUUBRAND™ 20700000
Only null left
Add to basket
Add to basket
The application-based VACUU·SELECT Controller with a touch screen display is easy-to-use, versatile, and efficient for all common vacuum processes in chemistry labs. Create unique applications with simple drag-and-drop editing, and detect boiling points for solvent evaporation.
  • Graphic user interface simplifies laboratory work
  • Predefined applications save time and guarantee reproducible results
  • Application editor allows easy customization
  • Integrated help function
  • Manual set-point-control for simple processes
  • Touch screen display can be operated even when wearing safety gloves
  • Ceramic vacuum sensor
  • Measuring Range: 0.1 to 1080 mbar (0.1 to 810) torr)
  • Measurement Principle: Ceramic Diaphragm (Alumina), Capacitive, Gas Independant, Absolute Pressure
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifications
Control Type VACUU·SELECT
Display Touchscreen
Limitations 0.1 to 1080 mbar
Quantity 1 Ea.
Sensors Ceramic Vacuum
Type Vacuum Controller