missing translation for 'onlineSavingsMsg'
Learn More

Brady™ V4500 Barcode Scanner USB Cable

Product Code. 30057748
Click to view available options
Length (English):
3.28 ft.
9.19 ft.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30057748

Brand: Brady™ 177237

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

V4500 Barcode Scanner accessories to ensure uninterrupted productivity.

Cable - USB Cable for V4500 Barcode Scanner

Specifications

Description USB to RJ50-1 Cable for V4500 Barcode Reader
Weight (Metric) 0.06 kg
Weight (English) 0.132 lb.
For Use With (Application) Asset Tracking, General Identification, Barcode Labelling, Circuit Board Labelling, Component and Equipment Labelling, Food and Beverage Process Labelling, Inventory and Inspection Labelling
For Use With (Equipment) V1300 Cradle (177235), V1300 Cradle Kit (177232), V4500 Barcode Scanner Kit (177229)
Length (English) 3.28 ft.
Type Cable
Alternate Part Number BRA-USB-RJ50-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.