Learn More
3M™ V4 Series Replacement Visor
Click to view available options
For Use With (Equipment):
V4C visor
V4D visor
V4F visor
Lens Material:
Acetate
Polycarbonate
Steel Mesh
Unit Size:
Pack of 10
Descrizione
- Easy to replace, visor snaps into place
- For use with 3M™ V4D Faceshield
- Replacement clear acetate visor offering good chemical resistant properties
- Acetate visor has good optical properties and an anti-fog coating
- Replacement clear polycarbonate visor offering impact and scratch resistance properties
- Replacement steel mesh visor with a black matt finish providing protection from flying chips and splinters
Specifica
Specifica
| Certifications/Compliance | CE Approved, EN1731 |
| Lens Tint | Clear |
| Product Type | V4 Series Replacement Visor |
| For Use With (Equipment) | V4D visor |
| Lens Material | Acetate |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto