Learn More
Uvex™ 9169 Super OTG Safety Spectacles
Product Code. 15325912 Shop All Uvex Products
Change view
Click to view available options
Lens Coating:
UV-treated
Lens Tint:
Clear
Frame Color:
Black
Unit Size:
Each
Product Code. Lens Coating Lens Tint Frame Color Unit Size
15325912 UV-treated Clear Black Each
1 options
Product Code. 15325912 Supplier Uvex™ Supplier No. 9169080
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Super OTG safety spectacles are very comfortable, while also providing excellent eye area protection. They will fit perfectly over conventional prescription lenses, making them ideal for visitors.
  • Extremely comfortable safety spectacles that offer excellent eye area protection
  • Designed to fit over presciption lenses, making these spectacles ideal for visitors
  • Hingeless arms with adjustable, soft arm ends for additional comfort
  • Uvex™ Supravision Sapphire coating is extremely scratch-resistant and chemical-resistant on both sides
  • Uvex™ Supravision Excellence coating features an anti-fog inside and an extremely scratch-resistant, chemical-resistant outside
  • Certified to EN 166 (personal eye protection) and EN 170 (UV filters)
  • Unrestricted peripheral vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 170
Lens Type Supravision Sapphire
Lens Coating UV-treated
Description Clear lens polycarbonate overspectacle
Lens Tint Clear
Frame Color Black
Temple Style Hingeless sidearms, Flexible ear pieces
Lens Material Polycarbonate
UV Protection UV400
Frame Type W 166 F CE