Learn More
Uvex™ i-vo Fog-Free Safety Glasses
Product Code. 15365872 Shop All Uvex Products
Click to view available options
:
Lens Coating: Uvex™ supravision excellence
Product Code. 15365872
Brand: Uvex™ 9160268
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Scratch-resistant and fog-resistant safety glasses
  • Eye-volution in eye protection with Duo Component Technology
  • Strap pitch and length adjustment, quattroflex strap, indirect ventilation, quick and easy disc change
  • Frame color blue / orange, W 166 FT CE
  • Disc brown / sun protection UV 400, 5-2,5 W 1 FTKN CE
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 172
    Lens Type Uvex™ supravision excellence
    Lens Coating Anti-fog, Scratch-resistant
    Description Strap glasses plastic
    Lens Tint Brown
    Frame Color Blue and orange