missing translation for 'onlineSavingsMsg'
Learn More

UTP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen UTP3
Concentration 0.24 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898714
View Documents
Proteintech
18886-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888714
View Documents
Proteintech
18886-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mediates apoptosis and actin stress fiber dissolution.
Specifications

Specifications

UTP3
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Rat, Mouse
Q6AXX4, Q9JI13, Q9NQZ2
305258, 57050, 65961
UTP3 Fusion Protein Ag13454
Primary
-20°C
UTP3
0.24 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CRL1, CRLZ1, Disrupter of silencing SAS10, SAS10, UTP3, UTP3 homolog
Utp3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.