missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18886-1-AP-20UL
This item is not returnable.
View return policy
Description
Mediates apoptosis and actin stress fiber dissolution.Specifications
| UTP3 | |
| Polyclonal | |
| Unconjugated | |
| UTP3 | |
| CRL1, CRLZ1, Disrupter of silencing SAS10, SAS10, UTP3, UTP3 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305258, 57050, 65961 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AXX4, Q9JI13, Q9NQZ2 | |
| Utp3 | |
| UTP3 Fusion Protein Ag13454 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction