missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | UTP20 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18294103
|
Novus Biologicals
NBP2-58128 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601208
|
Novus Biologicals
NBP2-58128-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UTP20 Polyclonal specifically detects UTP20 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| UTP20 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| Down-regulated in metastasis protein, DRIMdown regulated in metastasis, NNP73, Novel nucleolar protein 73, Protein Key-1A6, small subunit processome component 20 homolog, UTP20, small subunit (SSU) processome component, homolog (yeast) | |
| UTP20 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 27340 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FETLSDFESGLKYITDVVKLNAFDQRHLDDINFDVRFETFQTITSYIKEMQIVDVNYLIPVMHNCFYNLELGDMSLSDNASMC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title