missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP14A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | UTP14A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UTP14A Polyclonal specifically detects UTP14A in Human samples. It is validated for Western Blot.Specifications
| UTP14A | |
| Polyclonal | |
| Rabbit | |
| Q9BVJ6 | |
| 10813 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Antigen NY-CO-16, dJ537K23.3, FLJ37089, NYCO16, NY-CO-16, SDCCAG16, Serologically defined colon cancer antigen 16KIAA0266, U3 small nucleolar RNA-associated protein 14 homolog A, UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast) | |
| UTP14A | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title