missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP14A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56964
This item is not returnable.
View return policy
Description
UTP14A Polyclonal specifically detects UTP14A in Human samples. It is validated for Western Blot.
Specifications
| UTP14A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Antigen NY-CO-16, dJ537K23.3, FLJ37089, NYCO16, NY-CO-16, SDCCAG16, Serologically defined colon cancer antigen 16KIAA0266, U3 small nucleolar RNA-associated protein 14 homolog A, UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10813 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BVJ6 | |
| UTP14A | |
| Synthetic peptides corresponding to UTP14A (UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast)) The peptide sequence was selected from the N terminal of UTP14A. Peptide sequence KWDPVVLKNRQAEQLVFPLEKEEPAIAPIEHVLSGWKARTPLEQEIFNLL The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Equine: 92%; Chicken: 84%; Canine: 78%; Mouse: 78%; Rat: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction