missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP9Y Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £274.00
Specifications
| Antigen | USP9Y |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18354136
|
Novus Biologicals
NBP3-16073-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309456
|
Novus Biologicals
NBP3-16073-100UL |
100 μg |
£274.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP9Y Polyclonal antibody specifically detects USP9Y in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| USP9Y | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Deubiquitinating enzyme FAF-Y, DFFRYFLJ33043, EC 3.1.2.15, EC 3.4.19.12, Fat facets protein-related, Y-linked, probable ubiquitin carboxyl-terminal hydrolase FAF-Y, ubiquitin specific peptidase 9, Y-linked, ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-Y, Ubiquitin thiolesterase FAF-Y, ubiquitin-specific processing protease FAF-Y, Ubiquitin-specific protease 9, Y chromosome, Ubiquitin-specific-processing protease FAF-Y | |
| A synthetic peptide corresponding to a sequence within amino acids 2456-2555 of human USP9Y (NP_004645.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 8287 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title