missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP9Y Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16073-100UL
This item is not returnable.
View return policy
Description
USP9Y Polyclonal antibody specifically detects USP9Y in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| USP9Y | |
| Polyclonal | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Deubiquitinating enzyme FAF-Y, DFFRYFLJ33043, EC 3.1.2.15, EC 3.4.19.12, Fat facets protein-related, Y-linked, probable ubiquitin carboxyl-terminal hydrolase FAF-Y, ubiquitin specific peptidase 9, Y-linked, ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-Y, Ubiquitin thiolesterase FAF-Y, ubiquitin-specific processing protease FAF-Y, Ubiquitin-specific protease 9, Y chromosome, Ubiquitin-specific-processing protease FAF-Y | |
| A synthetic peptide corresponding to a sequence within amino acids 2456-2555 of human USP9Y (NP_004645.2). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8287 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction