missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | USP38 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877734
|
Proteintech
17767-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867734
|
Proteintech
17767-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Deubiquitinating enzyme exhibiting a preference towards ′Lys-63′-linked Ubiquitin chains.Specifications
USP38 | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q8BW70, Q8NB14 | |
307764, 74841, 84640 | |
USP38 Fusion Protein Ag12172 | |
Primary | |
-20°C | |
USP38 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 38, HP43.8KD, KIAA1891, Ubiquitin thiolesterase 38, USP38 | |
Usp38 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |