missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17767-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Deubiquitinating enzyme exhibiting a preference towards ′Lys-63′-linked Ubiquitin chains.Specifications
USP38 | |
Polyclonal | |
Unconjugated | |
USP38 | |
Deubiquitinating enzyme 38, HP43.8KD, KIAA1891, Ubiquitin thiolesterase 38, USP38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
307764, 74841, 84640 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BW70, Q8NB14 | |
Usp38 | |
USP38 Fusion Protein Ag12172 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |