missing translation for 'onlineSavingsMsg'
Learn More

USP26 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £366.00

Specifications

Antigen USP26
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18638240
View Documents
Novus Biologicals
NBP2-93692-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18675800
View Documents
Novus Biologicals
NBP2-93692-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

USP26 Polyclonal antibody specifically detects USP26 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

USP26
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cell Biology
PBS (pH 7.3), 50% glycerol
83844
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100
Polyclonal
Purified
RUO
Human, Mouse, Rat
Deubiquitinating enzyme 26, EC 3.4.19.12, MGC120066, MGC120067, MGC120068, ubiquitin carboxyl-terminal hydrolase 26, ubiquitin specific peptidase 26, ubiquitin specific protease 26, ubiquitin thioesterase 26, Ubiquitin thiolesterase 26, ubiquitin-specific processing protease 26, Ubiquitin-specific-processing protease 26
Recombinant fusion protein containing a sequence corresponding to amino acids 664-913 of human USP26 (NP_114113.1). TSLCQFHKAGGKPASSPGTPLSKVDFQTVPENPKRKKYVKTSKFVAFDRIINPTKDLYEDKNIRIPERFQKVSEQTQQCDGMRICEQAPQQALPQSFPKPGTQGHTKNLLRPTKLNLQKSNRNSLLALGSNKNPRNKDILDKIKSKAKETKRNDDKGDHTYRLISVVSHLGKTLKSGHYICDAYDFEKQIWFTYDDMRVLGIQEAQMQEDRRCTGYIFFYMHNEIFEEMLKREENAQLNSKEVEETLQKE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.