missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP26 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93692-0.02ml
This item is not returnable.
View return policy
Description
USP26 Polyclonal antibody specifically detects USP26 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| USP26 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Deubiquitinating enzyme 26, EC 3.4.19.12, MGC120066, MGC120067, MGC120068, ubiquitin carboxyl-terminal hydrolase 26, ubiquitin specific peptidase 26, ubiquitin specific protease 26, ubiquitin thioesterase 26, Ubiquitin thiolesterase 26, ubiquitin-specific processing protease 26, Ubiquitin-specific-processing protease 26 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 664-913 of human USP26 (NP_114113.1). TSLCQFHKAGGKPASSPGTPLSKVDFQTVPENPKRKKYVKTSKFVAFDRIINPTKDLYEDKNIRIPERFQKVSEQTQQCDGMRICEQAPQQALPQSFPKPGTQGHTKNLLRPTKLNLQKSNRNSLLALGSNKNPRNKDILDKIKSKAKETKRNDDKGDHTYRLISVVSHLGKTLKSGHYICDAYDFEKQIWFTYDDMRVLGIQEAQMQEDRRCTGYIFFYMHNEIFEEMLKREENAQLNSKEVEETLQKE | |
| 0.02 mL | |
| Cell Biology | |
| 83844 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction