missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | USP21 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18442131
|
Novus Biologicals
NBP1-86871-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18269557
|
Novus Biologicals
NBP1-86871 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP21 Polyclonal specifically detects USP21 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| USP21 | |
| Unconjugated | |
| RUO | |
| Human | |
| Deubiquitinating enzyme 21, EC 3.1.2.15, EC 3.4.19.12, MGC3394, ubiquitin carboxyl-terminal hydrolase 21, ubiquitin specific peptidase 21, ubiquitin specific protease 21, ubiquitin thioesterase 21, Ubiquitin thiolesterase 21, ubiquitin-specific processing protease 21, ubiquitin-specific protease 16, Ubiquitin-specific-processing protease 21, USP16, USP23NEDD8-specific protease | |
| USP21 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 27005 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CEAVNPTRFRAVFQKYVPSFSGYSQQDAQEFLKLLMERLHLEINRRGRRAPPILANGPVPSPPRRGGALLEEPELSDDDRANLMWKRYLEREDSKIVDLF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title