missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86871-25ul
This item is not returnable.
View return policy
Description
USP21 Polyclonal specifically detects USP21 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| USP21 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Deubiquitinating enzyme 21, EC 3.1.2.15, EC 3.4.19.12, MGC3394, ubiquitin carboxyl-terminal hydrolase 21, ubiquitin specific peptidase 21, ubiquitin specific protease 21, ubiquitin thioesterase 21, Ubiquitin thiolesterase 21, ubiquitin-specific processing protease 21, ubiquitin-specific protease 16, Ubiquitin-specific-processing protease 21, USP16, USP23NEDD8-specific protease | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| USP21 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CEAVNPTRFRAVFQKYVPSFSGYSQQDAQEFLKLLMERLHLEINRRGRRAPPILANGPVPSPPRRGGALLEEPELSDDDRANLMWKRYLEREDSKIVDLF | |
| 25 μL | |
| Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway | |
| 27005 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction