missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | USP13 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.25-1.0 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159841
|
Novus Biologicals
NBP2-24575SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108711
|
Novus Biologicals
NBP2-24575 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP13 Polyclonal specifically detects USP13 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| USP13 | |
| Western Blot 0.25-1.0 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Bovine, Canine, Equine, Primate | |
| Q92995 | |
| 8975 | |
| A portion of amino acids 500-550 of human USP13 was used as the immunogen for the antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13 | |
| USP13 | |
| IgG | |
| Affinity Purified | |
| This antibody is reactive to Chimpanzee, Cow, Dog, Horse, Human, Mouse, Rhesus Monkey. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title