missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24575
This item is not returnable.
View return policy
Description
USP13 Polyclonal specifically detects USP13 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| USP13 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8975 | |
| Human, Mouse, Bovine, Canine, Equine, Primate | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.25-1.0 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Q92995 | |
| USP13 | |
| A portion of amino acids 500-550 of human USP13 was used as the immunogen for the antibody. | |
| 0.1 mg | |
| Primary | |
| This antibody is reactive to Chimpanzee, Cow, Dog, Horse, Human, Mouse, Rhesus Monkey. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction