Learn More
Invitrogen™ USH1C Monoclonal Antibody (K06328_5A8), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | USH1C |
|---|---|
| Clone | K06328_5A8 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30210913
|
Invitrogen™
MA555966 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30203391
|
Invitrogen™
MA556346 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RERLAEARQR ELQRQELLMQ KRLAMESNKI LQEQQEMERQ RRKEIAQKAA EENERYRKEM EQIVEEEEKF KKQWEEDWGS KEQLLLPKTI TAEVHPVPLR KPKYDQGVEP ELEPADDLDG GTEEQGEQDF RKYEEGFDPY SMFTPEQIMG KDVRLLRIKK EGSLDLALEG GVDSPIGKVV VSAVYERGAA ERHGGIVKGD EIMAINGKIV TDYTLAEAEA ALQKAWNQGG DWIDLVVAVC PPKEYDDELT
USH1C Monoclonal antibody specifically detects USH1C in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| USH1C | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2010016F01Rik; AIE75; AIE-75; Antigen NY-CO-38/NY-CO-37; autoimmune enteropathy-related antigen AIE-75; DFNB18; DFNB18A; harmonin; harmonin a1; harmonin pseudogene; hypothetical protein LOC530709; hypothetical protein LOC564412; NY-CO-37; NY-CO-38; PDZ domain-containing protein; PDZ-45; PDZ73; PDZ-73; PDZ-73 protein; PDZ-73/NY-CO-38; PDZD7C; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; USH1 protein network component harmonin; Ush1c; ush1cpst; Usher syndrome 1C; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; usher syndrome type-1C protein; Usher syndrome type-1C protein homolog; zgc:136806 | |
| Recombinant human USH1C (immunogen range is 301-550). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| USH1C |
| K06328_5A8 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y6N9 | |
| 10083 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.