Learn More
Invitrogen™ USH1C Monoclonal Antibody (K06328_5A8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555966
Description
Sequence of this protein is as follows: RERLAEARQR ELQRQELLMQ KRLAMESNKI LQEQQEMERQ RRKEIAQKAA EENERYRKEM EQIVEEEEKF KKQWEEDWGS KEQLLLPKTI TAEVHPVPLR KPKYDQGVEP ELEPADDLDG GTEEQGEQDF RKYEEGFDPY SMFTPEQIMG KDVRLLRIKK EGSLDLALEG GVDSPIGKVV VSAVYERGAA ERHGGIVKGD EIMAINGKIV TDYTLAEAEA ALQKAWNQGG DWIDLVVAVC PPKEYDDELT
This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and non-syndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| USH1C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9Y6N9 | |
| USH1C | |
| Recombinant human USH1C (immunogen range is 301-550). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K06328_5A8 | |
| Unconjugated | |
| USH1C | |
| 2010016F01Rik; AIE75; AIE-75; Antigen NY-CO-38/NY-CO-37; autoimmune enteropathy-related antigen AIE-75; DFNB18; DFNB18A; harmonin; harmonin a1; harmonin pseudogene; hypothetical protein LOC530709; hypothetical protein LOC564412; NY-CO-37; NY-CO-38; PDZ domain-containing protein; PDZ-45; PDZ73; PDZ-73; PDZ-73 protein; PDZ-73/NY-CO-38; PDZD7C; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; USH1 protein network component harmonin; Ush1c; ush1cpst; Usher syndrome 1C; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; usher syndrome type-1C protein; Usher syndrome type-1C protein homolog; zgc:136806 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10083 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.