missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Urapidil hydrochloride, 98+%

Product Code. 15435479
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435479 500 mg 500mg
15415479 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435479 Supplier Thermo Scientific Chemicals Supplier No. J63219.ME
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist | CAS: 64887-14-5 | C20H29N5O3·HCl

alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64887-14-5
Molecular Formula C20H29N5O3·HCl
Synonym 6-(3-[4-(2-Methoxypenyl)-1-piperaziny]propylamino)-1,3-dimethyluracil hydrochloride

Specifications

Quantity 500 mg
Description White
Format Powder
For Use With (Application) alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist
Formula Weight 423.93g/mol
Product Type Signal Transduction Reagent
Form Powder
Assay ≥98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.