missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uracil-6-carboxaldehyde monohydrate, 98%
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
Uracil-6-carboxaldehyde monohydrate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Uracil-6-carboxaldehyde monohydrate is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Uracil-6-carboxaldehyde monohydrate is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Specifications
| Quantity | 250 mg |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 158.11 (140.10 Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | Uracil-6-carboxaldehyde monohydrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction